Sino Biological™ ICOSLG (ICOSL) Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human ICOSLG (NP_056074.1) extracellular domain (Met 1-Ser 258) fused to the polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 11559H08H50

 View more versions of this product

Catalog No. 11-559-H08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 28kDa
    Purity ≥95% by SDS-PAGE
    Quantity 50μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Other Proteins
    Endotoxin Concentration <1 EU/μg
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Gene Alias B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, KIAA0653, LICOS
    Common Name Inducible T-cell co-stimulator ligand
    Gene Symbol ICOSLG (ICOSL)
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family CD Proteins
    Expression System Mammalian

    Inducible T-cell co-stimulator ligand (ICOSLG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized human ICOSLG at 1µg/mL (100µL/well) can bind human ICOS with a linear range of 1.6-200ng/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit