Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ IFNG Recombinant Mouse Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse IFNG (P01580) (Met 1-Cys 155) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Invitrogen 50709M02H25

 View more versions of this product

Catalog No. 50-709-M02H25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Specifications

Description

Interferon gamma (IFNG) recombinant mouse protein is supplied as a lyophilized powder
  • Activity: Measured in anti-viral assays using L929 cells infected with vesicular stomatitisvirus (VSV); the ED50 for this effect is typically 20–80ng/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween® 80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
15978
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Interferon gamma
Assay Standard
Mammalian
Lyophilized
42.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
IFN-g, Ifg
IFNG
Cytokines & Receptors
IgG1 Fc Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit