Sino Biological™ IGF1 Recombinant Human Protein, His.NusA Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF1 isoform 1 (P05019-1) (Gly 49-Ala 118) fused to the polyhistidine-NusA tag at the N-terminus

Manufacturer: Invitrogen 10598H24E5

 View more versions of this product

Catalog No. 10-598-H24E5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Common Name Insulin-like growth factor 1 (somatomedin C)
    Gene Symbol IGF1
    Protein Tag His Tag/NusA Tag
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 64.8kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Structural Form Full Length/Recombinant
    Protein Subtype IGFs (Insulin-Like Growth Factors)
    Gene Alias IGF-I, IGF1A, IGFI
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors
    Expression System E. coli
    Format Lyophilized

    Insulin-like growth factor 1 (IGF1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized human IGF1 at 10µg/mL (100µL/well) can bind biotinylated human IGFBP3-His, the EC50 of biotinylated human IGFBP3-His is 24ng/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit