Sino Biological™ IGFBP4 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human IGFBP4 (NP_001543.2) precursor (Met 1-Glu 258) fused to a C-terminal polyhistidine tag

Manufacturer: Invitrogen 50250M08H250

 View more versions of this product

Catalog No. 50-LM0-8HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

    Specifications

    Molecular Weight 27.2kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 50μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype IGFs (Insulin-Like Growth Factors)|Growth Factor Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Alias Igfbp4, RP23-333D2.6, AI875747, Deb2, IGFBP-4
    Common Name Insulin-like growth factor binding protein 4
    Gene Symbol IGFBP4
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Insulin-Like Growth Factor Binding Protein 4 (IGFBP4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

    • Activity: Measured in a serum-free cell proliferation assay using MCF7 human breast adenocarcinoma cells (Karey, K.P. et al. 1988, Cancer Research 48: 4083); the ED50 is typically 5-10µg/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit