Sino Biological™ IL18BP Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the precursor (Met 1-Ala 193) of mouse IL18BP (NP_034661.1) fused to a polyhistide tag at the C-terminus

Manufacturer: Invitrogen 50206M08H25

 View more versions of this product

Catalog No. 50-206-M08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 16068
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Recombinant/Ligand Binding Domain
    Protein Tag His Tag
    Validated Application Assay Standard
    Expression System Mammalian
    Format Lyophilized
    Molecular Weight 19.4kDa
    Protein Subtype Interleukins
    Endotoxin Concentration <1 EU/μg
    Gene Alias Il18bp, IL-18BP, Igifbp, MC54L
    Common Name Interleukin 18 binding protein
    Gene Symbol IL8BP
    Protein Family Cytokines & Receptors
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

    Interleukin 18 binding protein (IL18BP), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse IL18BP-His at 1µg/well can bind biotinylated cynomolgus IL18 with a linear range of 0.16-4.0µg/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit