Sino Biological™ IL18R1 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 329) of human IL18R1 (NP_003846.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 11102H08H250

 View more versions of this product

Catalog No. 11-102-H08HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 8809
    Molecular Weight 37.2kDa
    Quantity 5 x 50μg
    Structural Form Truncated/Recombinant
    Protein Subtype Interleukins|Cytokine Receptors
    Common Name Interleukin 18 receptor 1
    Gene Symbol IL18R1
    Protein Tag His Tag
    Expression System Mammalian
    Purity ≥90% by SDS-PAGE
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Gene Alias IL18R1, CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
    Validated Application Assay Standard
    Protein Family Cytokines & Receptors
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Interleukin 18 Receptor 1 (IL18R1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

    • Activity: This recombinant protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit