Sino Biological™ IL1A Recombinant Mouse Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse IL1A (NP_034684.2) (Ser 115-Ser 270), with an initial Met at the N-terminus

Manufacturer: Invitrogen 50114MNAE50

 View more versions of this product

Catalog No. 50-114-MNAE50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

    Specifications

    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Common Name Interleukin 1 alpha
    Protein Family Cytokines & Receptors
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 17kDa
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Protein Subtype Interleukins
    Gene Alias RP23-160G19.8, Il-1a
    Gene Symbol IL1A
    Protein Tag Untagged
    Validated Application Assay Standard
    Expression System E. coli
    Format Lyophilized

    Interleukin 1 alpha (IL1A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

    • Activity: 1. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells, the ED50 for this effect is typically 2-10pg/mL; 2. Measured by its binding ability in a functional ELISA; immobilized mouse IL1A at 10µg/mL (100µL/well) can bind mouse IL1R1, the EC50 of mouse IL1R1 is 0.13µg/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit