Sino Biological™ IL1F5 Recombinant Mouse Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse IL1F5 (NP_062324.2) (Met 2-Asp 156)

Manufacturer: Invitrogen 50213MNAE5

 View more versions of this product

Catalog No. 50-213-MNAE5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of protein in suspension buffer

    Specifications

    Gene ID (Entrez) 54450
    Molecular Weight 17kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Structural Form Full Length/Recombinant
    Protein Subtype Interleukins
    Common Name Interleukin 1 family member 5 (delta)
    Validated Application Assay Standard
    Expression System E. coli
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Purification Method PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
    Endotoxin Concentration <1 EU/μg
    Gene Alias RP23-176J12.6, AI413231, Fil1delta, Il-1h3, Il1hy1
    Gene Symbol IL1F5
    Protein Family Cytokines & Receptors

    Interleukin 1 Family Member 5 (Delta) (IL1F5), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: The activity of this protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit