Sino Biological™ IL6ST Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse extracellular domain (Met 1-Glu 617) of IL6ST (NP_034690.3) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50135M08H5

 View more versions of this product

Catalog No. 50-135-M08H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 16195
    Purity ≥90% by SDS-PAGE
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Interleukins|Cytokine Receptors
    Protein Tag His Tag
    Protein Family Cytokines & Receptors/CD Proteins
    Expression System Mammalian
    Molecular Weight 68kDa
    Quantity 5μg
    Structural Form Recombinant
    Endotoxin Concentration <1 EU/μg
    Gene Alias IL6ST, 5133400A03Rik, AA389424, BB405851, CD130, D13Ertd699e, gp130
    Common Name Interleukin 6 signal transducer
    Gene Symbol IL6ST
    Validated Application Assay Standard
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Interleukin 6 signal transducer (IL6ST), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

    • Activity: 1. Measured by its ability to bind mouse IL11RA in a functional ELISA; 2. Measured by its ability to inhibit the IL6R enhancement of IL6 activity on M1 mouse myeloid leukemia cells (Saito, T. et al. 1991, J. Immunol. 147:168), the ED50 for this effect is typically 0.2-0.8µg/mL in the presence of 50ng/mL recombinant human IL6sR and 100ng/mL recombinant human IL6
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit