Sino Biological™ IL7R (CD127) Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 236) of human IL7R (NP_002176.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 10975H08H50

 View more versions of this product

Catalog No. 10-975-H08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 3575
    Purity ≥90% by SDS-PAGE
    Protein Subtype Interleukins|Cytokine Receptors
    Endotoxin Concentration <1 EU/μg
    Common Name Interleukin 7 receptor
    Gene Symbol IL7R
    Validated Application Assay Standard
    Protein Family Cytokines & Receptors/CD Proteins
    Expression System Mammalian
    Format Lyophilized
    Molecular Weight 26.3kDa
    Quantity 50μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant
    Gene Alias IL7R, CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
    Protein Tag His Tag
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

    Interleukin 7 receptor (IL7R/CD127), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein-protein interaction assays, protein structure analysis, as immunogen, protein standard, and other research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL7R-His at 1µg/well can bind biotinylated mouse TSLP-His with a linear range of 1.25-10.0µg/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit