Sino Biological™ INSR, Short Isoform, Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 944) of human INSR short isoform (NP_001073285.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 11086H08H50

 View more versions of this product

Catalog No. 11-086-H08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 3643
    Molecular Weight 106.0kDa
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Protein Subtype Hormone Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Alias CD220, HHF5, INSR
    Common Name Insulin receptor, short isoform
    Gene Symbol INSR
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Hormones & Receptors
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant/Cell Surface Protein
    KinaseFamily Insulin & IGF Receptor Kinase Family
    Format Lyophilized

    Insulin receptor (INSR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; when 1µg/mL of biotinylated human insulin is immobilized onto a streptavidin coated plate, it can bind human insulin receptor (INSR) with a linear range of 0.3-40µg/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit