Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ KITL Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse KITL (P20826-1) (Met 1-Ala 189) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 50487M08B250

 View more versions of this product

Catalog No. 50-487-M08BL


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Specifications

Description

KIT ligand (KITL), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized mouse KITL at 2µg/mL (100µL/well) can bind mouse KIT with a linear ranger of 1.28-32ng/mL; 2. Measured in a cell proliferation assay using TF-1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323), the EC50 for this effect is typically 0.5-5ng/mL
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
19.7kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Kit ligand
Assay Standard
Insect
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Growth Factors
Clo, Con, Gb, Kitlg, mgf, SCF, SF, SLF, Sl, Steel, c ontrasted
KITL
Growth Factors & Receptors
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit