Sino Biological™ KITL Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse KITL (P20826-1) (Met 1-Ala 189) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50487M08B5

 View more versions of this product

Catalog No. 50-487-M08B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

    Specifications

    Molecular Weight 19.7kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Protein Subtype Other Growth Factors
    Endotoxin Concentration <1 EU/μg
    Gene Alias Clo, Con, Gb, Kitlg, mgf, SCF, SF, SLF, Sl, Steel, c ontrasted
    Common Name Kit ligand
    Gene Symbol KITL
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors
    Expression System Insect
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    KIT ligand (KITL), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

    • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized mouse KITL at 2µg/mL (100µL/well) can bind mouse KIT with a linear ranger of 1.28-32ng/mL; 2. Measured in a cell proliferation assay using TF-1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323), the EC50 for this effect is typically 0.5-5ng/mL
    • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit