Sino Biological™ LAIR2 Recombinant Human Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human LAIR2 (NP_002279.2) (Met 1-Pro 152)

Manufacturer: Invitrogen 10868HNAH25

 View more versions of this product

Catalog No. 10-868-HNAH25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 14.1kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Other Proteins
    Endotoxin Concentration <1 EU/μg
    Common Name Leukocyte-associated immunoglobulin-like receptor 2
    Gene Symbol LAIR2
    Protein Tag Untagged
    Validated Application Assay Standard
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Gene Alias LAIR2, XXbac-BCX535A19.6, CD306, mgC71634
    Protein Family CD Proteins

    Leukocyte-Associated Immunoglobulin-Like Receptor 2 (LAIR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

    • Activity: Measured by the ability of the immobilized protein to support the adhesion of HT-29 human colon adenocarcinoma cells; when 5 x 104 cells/well are added to recombiannt human LAIR2 coated plates (50µg/mL with 100µL/well), approximately 30% will adhere after 30 minutes at 37°C (optimal concentration depends on cell type as well as the application or research objectives)
    • Formulation: Lyophilized in 50mM Tris, 0.7M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit