Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ LCN2 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse LCN2 precursor (NP_032517.1) (Met 1-Asn 200) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 50060M08H250

 View more versions of this product

Catalog No. 50-060-M08HL


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Specifications

Description

Lipocalin 2 (LCN2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing functional assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind Iron(III) dihydroxybenzoic acid (Fe(DHBA)3); the binding of Fe(DHBA)3 results in the quenching of Trp fluorescence in recombinant mouse Lipocalin 2 (LCN2); recombinant mouse LCN2 can bind >1.0µM of Fe(DHBA)3
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
22.3kDa
5 x 50μg
Other Proteins
Lcn2, RP23-161B9.11, 24p3, AW212229, Sip24
LCN2
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Lipocalin 2
Assay Standard
Mammalian
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit