Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ mMP9 Recombinant Rat Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the ratmMP9 (EDL96479.1) (Met 1-Pro 708) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 80049R08H5

 View more versions of this product

Catalog No. 80-049-R08H5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Matrix Metallopeptidase 9 (MMP9), recombinant rat protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (R&D Systems Cat. No. ES010); the specific activity is >1000pmol/min./µg
  • Formulation: 50mM MES, 100mM NaCl, 1mM CaCl2, 0.2µg/µL BSA, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
77.8kDa
5μg
Proteases
Mmp9
MMP9
Mammalian
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Matrix metallopeptidase 9
Assay Standard
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit