Sino Biological™ Neuraminidase (NA) Recombinant Influenza A Virus Protein Subtype H3N2 (A/Babol/36/2005), His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the Influenza A H3N2 virus (A/Babol/36/2005) Neuraminidase (ACN50232.1) (His 36-Pro 459) fused to a polyhistidine tag at the N-terminus

Manufacturer: Invitrogen 40017V07H5

 View more versions of this product

Catalog No. 40-017-V07H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Purification Method PAGE
    Purity ≥90% by SDS-PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
    Termination N-Terminal
    Structural Form Full Length/Recombinant
    Molecular Weight 49.6kDa
    Quantity 5μg
    Protein Subtype Viral Proteins
    Endotoxin Concentration <1 EU/μg
    Gene Alias NA
    Common Name Neuraminidase
    Gene Symbol NA
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Gene ID (Entrez) Unknown

    Neuraminidase (NA), recombinant Influenza A H3N2 ( A/Babol/36/2005 ) NA/Neuraminidase protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: The activity of this protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit