Sino Biological™ Neuraminidase (NA) Recombinant Viral Protein, N-His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of influenza A virus (A/California/04/2009 (H1N1)) neuraminidase (ACP41107.1) (His 36 -Lys 469) fused to a polyhistidine tag at the N-terminus

Manufacturer: Invitrogen 11058V07B5

Catalog No. 11-058-V07B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Purity ≥85% by SDS-PAGE
    Protein Subtype Viral Proteins
    Endotoxin Concentration <1 EU/μg
    Protein Family Other Proteins
    Expression System Insect
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 50kDa
    Purification Method PAGE
    Quantity 5μg
    Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Termination N-Terminal
    Structural Form Full Length/Recombinant
    Gene Alias NA
    Common Name Neuraminidase/NA Recombinant Influenza A Virus Protein subtype H1N1 (A/California/04 /2009)
    Gene Symbol NA/Neuraminidase
    Protein Tag His Tag
    Validated Application Assay Standard
    Format Frozen/Lyophilized

    NA Recombinant Influenza A Virus Protein subtype H1N1 (A/California/04 /2009) (NEURAMINIDASE), recombinant viral protein is supplied as a frozen protein. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its ability to cleave a fluorogenic substrate, 2'-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid, the specific activity is 800-1200pmole/min./µg
    • Formulation: 50mM Tris-Citrate, 0.3M NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: No reconstitution needed

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit