Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ NGF Recombinant Mouse Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse NGF (NP_001106168.1), residues 122 to 241, with an initial Met at the N-terminus

Manufacturer:  Invitrogen 50385MNAC5

 View more versions of this product

Catalog No. 50-385-MNAC5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Nerve growth factor (NGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells; the ED50 for this effect is 0.2-2.0ng/mL
  • Formulation: Lyophilized in 50mM Na acetate buffer, pH 5.5, 150mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
18049
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Neurotrophic Factors
Ngf, Ngfb
NGF
Growth Factors & Receptors
Untagged
Lyophilized
13.5kDa
5μg
Native
<1 EU/μg
Nerve growth factor
Assay Standard
Mammalian
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit