Sino Biological™ NGF Recombinant Mouse Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse NGF (NP_001106168.1), residues 122 to 241, with an initial Met at the N-terminus

Manufacturer: Invitrogen 50385MNAC50

 View more versions of this product

Catalog No. 50-385-MNAC50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 18049
    Purity ≥95% by SDS-PAGE
    Quantity 50μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Native
    Protein Subtype Neurotrophic Factors
    Endotoxin Concentration <1 EU/μg
    Gene Alias Ngf, Ngfb
    Common Name Nerve growth factor
    Gene Symbol NGF
    Protein Tag Untagged
    Protein Family Growth Factors & Receptors
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 13.5kDa
    Validated Application Assay Standard
    Format Lyophilized

    Nerve growth factor (NGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.

    • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells; the ED50 for this effect is 0.2-2.0ng/mL
    • Formulation: Lyophilized in 50mM Na acetate buffer, pH 5.5, 150mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit