Sino Biological™ PCSK9 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the full length of mouse PCSK9 (NP_705793.1) precursor (Met 1-Gln 694) fused to a C-terminal polyhistidine tag

Manufacturer: Invitrogen 50251M08H5

 View more versions of this product

Catalog No. 50-251-M08H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 72.6kDa
    Quantity 5μg
    Protein Subtype Proteases
    Common Name Proprotein convertase subtilisin/kexin type 9
    Protein Tag His Tag
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Purity ≥90% by SDS-PAGE
    Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Gene Alias RP23-159J7.1, AI415265, AI747682, FH3, HCHOLA3, mgC47409, Narc1
    Gene Symbol PCSK9
    Validated Application Assay Standard
    Format Frozen

    Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse PCSK9 at 10µg/mL (100µL/well) can bind biotinylated recombinant human LDLR; the EC50 of biotinylated human LDLR is 0.12µg/mL
    • Formulation: 15mM Tris, 90mM NaCl, 50% glycerol, pH 7.5
    • Reconstitution: No reconstitution needed

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit