Sino Biological™ PCSK9 Recombinant Rat Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the rat PCSK9 (NP_954862.2) (Met 1-Gln 691) fused to a polyhistidine tag at the N-terminus

Manufacturer: Invitrogen 80005R08H5

 View more versions of this product

Catalog No. 80-005-R08H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

    Specifications

    Molecular Weight 20kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Proteases
    Endotoxin Concentration <1 EU/μg
    Gene Alias NARC-1, Narc1, PC9, PCSK9
    Common Name Proprotein convertase subtilisin/kexin type 9
    Gene Symbol PCSK9
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Proprotein convertase subtilisin/kexin type 9 (PCSK9), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: 1. Measured by its ability to bind biotinylated human LDLR in a functional ELISA; 2. Measured by its ability to bind biotinylated mouse LDLR in a functional ELISA
    • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit