Sino Biological™ PDCD1LG2 Recombinant Mouse Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse PDCD1LG2 (Q9WUL5) (Met 1-Arg 219) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: Invitrogen 50804M02H250

 View more versions of this product

Catalog No. 50-804-M02HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

    Specifications

    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Gene Alias B7-DC, Btdc, F730015O22Rik, mgC124039, mgC124040, PD-L2B7-DC, Btdc, F730015O22Rik, PD-L2
    Common Name Programmed cell death 1 ligand 2
    Validated Application Assay Standard
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 49.5kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 50μg
    Protein Subtype Other Proteins
    Gene Symbol PDCD1LG2
    Protein Tag IgG1 Fc Tag
    Protein Family CD Proteins
    Format Lyophilized

    Programmed cell death 1 ligand 2 (PDCD1LG2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized human PDCD1-His (Mfr. No. 10377-H08H) at 10µg/mL (100µL/well) can bind mouse PDCD1LG2-Fc; the EC50 of mouse PDCD1LG2-Fc is 0.95µg/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit