Sino Biological™ PTPN2 Recombinant Mouse Protein, N-His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the amino acid (Met 1-Asn 314) of human PTPN2 (P17706-1) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Manufacturer: Invitrogen 50800M07B5

 View more versions of this product

Catalog No. 50-800-M07B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 38.7kDa
    Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Tyrosine Phosphatases
    Gene Symbol PTPN2
    Protein Tag His Tag
    Expression System Insect
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Purity ≥85% by SDS-PAGE
    Quantity 5μg
    Endotoxin Concentration <1 EU/μg
    Gene Alias AI325124, Ptpt, TC-PTP
    Common Name Protein tyrosine phosphatase, nonreceptor type 2
    Validated Application Assay Standard
    Format Frozen

    Protein Tyrosine Phosphatase Nonreceptor Type 2 (PTPN2), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor (988-998 phosphopeptide substrate, R&D Systems Cat. No. ES006); the specific activity is >15nM/min/µg
    • Formulation: 20mM Tris, 500mM NaCl, 0.2µg/µL BSA, pH 8.0, 20% glycerol, 0.1mM TCEP
    • Reconstitution: No reconstitution needed

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit