Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ PVR Recombinant Rat Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of rat PVR (NP_058772.2) (Met 1-Gly 352) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 80007R08H25

 View more versions of this product

Catalog No. 80-007-R08H25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Specifications

Description

Poliovirus receptor (PVR), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized rat PVR at 20µg/mL (100µL/well) can bind mouse CD226 with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
35.8kDa
5 x 5μg
Viral Host Cell Receptors
PVR, Taa1, Tage4
PVR
CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Poliovirus receptor
Assay Standard
Mammalian
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit