Sino Biological™ RBP4 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse RBP4 (NP_035385.1) (Met 1-Leu 201) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50170M08H5

 View more versions of this product

Catalog No. 50-170-M08H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

    Specifications

    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Validated Application Assay Standard
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 22.8kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Protein Subtype Other Proteins
    Endotoxin Concentration <1 EU/μg
    Gene Alias Rbp-4, Rbp4
    Common Name RBP4 retinol binding protein 4, plasma
    Gene Symbol RBP4
    Protein Tag His Tag
    Protein Family Other Proteins
    Expression System Mammalian
    Format Lyophilized

    Retinol binding protein 4, plasma (RBP4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its ability to bind all trans retinoic acid; the binding of retinoic acid results in the quenching of Trp fluorescence in RBP4; the 50% binding concentration (BC50) is >0.5µM
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit