Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ SCARB2 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse SCARB2 (NP_031670.1) (Met 1-Thr 432) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 50052M08H250

 View more versions of this product

Catalog No. 50-052-M08HL


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer

Specifications

Description

Scavenger Receptor Class B, Member 2 (SCARB2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris-Citrate, 300mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12492
PAGE
5 x 50μg
C-Terminal
<1 EU/μg
Scavenger receptor class B, member 2
Assay Standard
Mammalian
Lyophilized
47.8kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
Scarb2, 9330185J12Rik, Cd36l2, LGP85, LIMP-2, MLGP85
SCARB2
CD Proteins
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit