Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sino Biological™ SMYD3 Recombinant Human Protein, His Tag N-Flag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human SMYD3 isoform 1 (NP_001161212.1) (Met 1-Ser 428) fused to the flag tag at the N-terminus and the Histidine tag at the C-terminus

Manufacturer:  Thermo Scientific™ 11230H18H5

Catalog No. 11-230-H18H5


Add to cart

Description

Description

SET and MYND domain containing 3 (SMYD3), recombinant human protein is supplied frozen. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 25mM Tris, 100mM NaCl, 20% glycerol, 3mM DTT, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
64754
PAGE
5μg
Full Length/Recombinant
FLJ21080, KMT3E, mgC104324, ZMYND1, ZNFN3A1, bA74P14.1, SMYD3
SMYD3
Methylation & Demethylation Proteins
N-Flag C-His
Frozen
51.5kDa
≥85% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Set And Mynd Domain Containing 3
Assay Standard
Mammalian
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.