Sino Biological™ TNF (TNFA) Recombinant Canine Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the soluble form of canine TNFα (NP_001003244.4) (Val 77-Leu 233) fused to an initial Met at the N-terminus

Manufacturer: Invitrogen 70003DNAE25

 View more versions of this product

Catalog No. 70-003-DNAE25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Protein Subtype TNF (Tumor Necrosis Factor)
    Common Name Tumor necrosis factor
    Protein Tag Untagged
    Validated Application Assay Standard
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Frozen
    Molecular Weight 17.3kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Gene Alias TNFA, Ctnf
    Gene Symbol TNF
    Expression System E. coli

    Tumor Necrosis Factor (TNF), recombinant canine protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

    • Activity: Measured in a cytotoxicity assay using L-929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D; the ED50 is typically 0.2-1.2ng/mL
    • Formulation: 50mM Tris, 500mM NaCl, 20% glycerol, pH 8.0
    • Reconstitution: No reconstitution needed

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit