Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ TNF (TNFA) Recombinant Canine Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the soluble form of canine TNFα (NP_001003244.4) (Val 77-Leu 233) fused to an initial Met at the N-terminus

Manufacturer:  Invitrogen 70003DNAE5

 View more versions of this product

Catalog No. 70-003-DNAE5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Tumor Necrosis Factor (TNF), recombinant canine protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cytotoxicity assay using L-929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D; the ED50 is typically 0.2-1.2ng/mL
  • Formulation: 50mM Tris, 500mM NaCl, 20% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
17.3kDa
5μg
TNF (Tumor Necrosis Factor)
Tumor necrosis factor
Assay Standard
Untagged
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
TNFA, Ctnf
TNF
E. coli
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit