Sino Biological™ TNFRSF17 Recombinant Human Protein, hIgG1-Fc.His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TNFRSF17 (NP_001183.2) (Met 1-Ala 54) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus and a signal peptide at the N-terminus

Manufacturer: Invitrogen 10620H03H25

 View more versions of this product

Catalog No. 10-620-H03H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 34kDa
    Purity ≥85% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype TNF Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Alias TNFRSF17, BCM, BCMA, CD269
    Common Name Tumor necrosis factor receptor superfamily, member 17
    Gene Symbol TNFRSF17
    Protein Tag His Tag/IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Cytokines & Receptors/CD Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Tumor necrosis factor receptor superfamily, member 17 (TNFRSF17), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human TNFSF13B/BAFF at 1µg/mL (100µL/well) can bind human TNFRSF17, the EC50 of human TNFRSF17 is 0.07µg/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit