Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Sino Biological™ TNFRSF1A (CD120a) Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 212) of mouse TNFRSF1A (NP_ 035739.2) fused to a polyhistidine tag at the C-terminus
Manufacturer: Invitrogen 50496M08H25
 View more versions of this product
Catalog No. 50-496-M08H25
  N/A
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Related Products
Product Content Feedback