Sino Biological™ TNFRSF25 Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human TNFRSF25 (NP_003781.1) extracellular domain (Met 1-Gln 199) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: Invitrogen 12095H02H25

 View more versions of this product

Catalog No. 12-095-H02H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 8718
    Molecular Weight 46kDa
    Purity ≥90% by SDS-PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
    Endotoxin Concentration <1 EU/μg
    Gene Symbol TNFRSF25
    Protein Tag IgG1 Fc Tag
    Expression System Mammalian
    Format Lyophilized
    Purification Method PAGE
    Quantity 5 x 5μg
    Termination C-Terminal
    Structural Form Truncated/Recombinant
    Gene Alias RP4-650H14.2, APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR
    Common Name Tumor necrosis factor receptor superfamily, member 25
    Validated Application Assay Standard
    Protein Family Cytokines & Receptors
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

    Tumor Necrosis Factor Receptor Superfamily, Member 25 (TNFRSF25), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

    • Activity: The activity of this protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit