Sino Biological™ TNFSF9 Recombinant Mouse Protein, N-His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Arg 104-Glu 309) of the mouse TNFSF9 precursor (NP_033430.1) fused to a polyhistidine tag at the N-terminus

Manufacturer: Invitrogen 50067M07H25

 View more versions of this product

Catalog No. 50-067-M07H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

    Specifications

    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Protein Subtype TNF (Tumor Necrosis Factor)
    Endotoxin Concentration <1 EU/μg
    Gene Alias TNFSF9, 4-1BB-L, 4-1BBL, AI848817, Cd137l, Ly63l
    Common Name Tumor necrosis factor (ligand) superfamily, member 9
    Protein Tag His Tag
    Protein Family Cytokines & Receptors/CD Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 25kDa
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Gene Symbol TNFSF9
    Validated Application Assay Standard
    Format Lyophilized

    Tumor necrosis factor (ligand) superfamily, member 9 (TNFSF9), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse TNFSF9 at 20µg/mL (100µL/well) can bind human TNFSRF9 with a linear range of 15.6-500ng/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit