Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ TP53 (P53) Recombinant Cynomolgus Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the cynomolgus (Macaca fascicularis) P53 (E3U906) (Met 1-Asp 393)

Manufacturer:  Invitrogen 90001CNAE5

 View more versions of this product

Catalog No. 90-001-CNAE5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

Specifications

Description

Tumor protein p53 (P53), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind biotinylated human S100B in a functional ELISA; 2. Measured by its ability to bind biotinylated human RRM2B in a functional ELISA
  • Formulation: Lyophilized in 100mM Tris, 3% Glycerol, 4mM DTT, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
43.7kDa
5μg
Other Proteins
Cynomolgus Cellular Tumor Antigen p53
Assay Standard
E. coli
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
p53, TP53
TP53
Other Proteins
Untagged
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit