Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sino Biological™ TREM2 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain of human TREM2 (NP_061838.1) (Met 1-Ser 174) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Thermo Scientific™ 11084H08H5

Catalog No. 11-084-H08H5


Add to cart

Description

Description

Triggering Receptor Expressed on Myeloid Cells 2 (TREM2) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.Background: Triggering receptor expressed on myeloid cells 2 (TREM2) is a single Ig domain receptor. It is expressed on macrophages and dendritic cells but not on granulocytes or monocytes. TREM2 may play a role in chronic inflammations and may stimulate production of constitutive rather than inflammatory chemokines and cytokines. TREM2 forms a receptor signaling complex with TYROBP and triggers activation of the immune responses in macrophages and dendritic cells. It also associates with the signal adapter protein, DAP12, which has a cytoplasmic ITAM, leading to the subsequent activation of cytoplasmic tyrosine kinases. TREM2 is both required and sufficient for competent uptake of apoptotic neuronal cells. TREM2 and TREM2-L form a receptor-ligand pair connecting microglia with apoptotic neurons, directing removal of damaged cells to allow repair. Deficiency of the adapter protein DAP12 or its associated receptor TREM2 is associated with abnormal osteoclast development in humans. Defects in TREM2 are causes of PLOSL, also known as NHD. In addition, TREM2 signaling is also an important pathway to promote healing.
  • N-terminal Sequence Analysis: His 19
  • Activity: The activity of this protein has not been tested
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween™-80
  • Reconstitution: dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Manufactured by Sino Biological (China).

Specifications

Specifications

Assay Standard
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
54209
PAGE
5μg
C-Terminal
<1 EU/μg
Triggering receptor expressed on myeloid cells 2
His Tag
Mammalian
Other Proteins
Lyophilized
18.9kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
TREM-2, Trem2a, Trem2b, Trem2c, TREM2
TREM2
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.