Sino Biological™ UCHL1 (UCH-L1) Recombinant Mouse Protein, N-His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse UCHL1 (Q9R0P9) (Gln 2-Ala 223) fused to a polyhistide tag at the N-terminus

Manufacturer: Invitrogen 50690M07E25

 View more versions of this product

Catalog No. 50-690-M07E25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer Store at -80 °C.

    Specifications

    Gene ID (Entrez) 22223
    Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Common Name Ubiquitin carboxyl-terminal esterase L1/Ubiquitin Carboxy-terminal Hydrolase L1
    Expression System E. coli
    Molecular Weight 26.2kDa
    Purification Method PAGE
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Termination N-Terminal
    Structural Form Full Length/Recombinant
    Protein Subtype Proteases
    Gene Alias AW822034, C88048, PGP9.5, R75593, gad
    Gene Symbol UCHL1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Frozen/Lyophilized

    Ubiquitin Carboxy-terminal Hydrolase L1 (UCHL1), recombinant mouse protein is supplied frozen. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by the hydrolysis of UbiquitinAMC; the specific activity is >14,000pmole/min./µg
    • Formulation: 50mM Tris, 150mM NaCl, 20% Glycerol, pH 7.7, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: No reconstitution needed

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit