Sino Biological™ USP7 Recombinant Human Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the amino acid sequence (Lys 208-Glu 560) of human USP7 (NP_003461.2) fused to two additional aa (Gly & Pro) at the N-terminus

Manufacturer: Invitrogen 11681HNCB250

 View all options for this product

Catalog No. 11-681-HNCBL

  Check Availability
Add to Cart


For Research Use Only. Not for use in diagnostic procedures.


    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Gene ID (Entrez) 7874
    Molecular Weight 41kDa
    Purification Method PAGE
    Purity ≥95% by SDS-PAGE
    Quantity 5 x 50μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
    Structural Form Truncated/Recombinant
    Endotoxin Concentration <1 EU/μg
    Gene Alias HAUSP, TEF1
    Common Name Ubiquitin carboxyl-terminal hydrolase 7
    Gene Symbol USP7
    Validated Application Assay Standard
    Protein Family Proteases, Peptidases & Inhibitors
    Expression System Insect

    Ubiquitin Carboxyl-Terminal Hydrolase 7 (USP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: The activity of this protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products

Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit