Sino Biological™ VNN1 Recombinant Mouse Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mouse VNN1 (Q9Z0K8) (Met 1-Ser 487) without the pro peptides

Manufacturer: Invitrogen 50952M02H25

 View more versions of this product

Catalog No. 50-952-M02H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer

    Specifications

    Gene ID (Entrez) 22361
    Molecular Weight 79kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Termination C-Terminal
    Validated Application Assay Standard
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Purification Method PAGE
    Structural Form Full Length/Recombinant
    Endotoxin Concentration <1 EU/μg
    Gene Alias V-1
    Common Name Vanin 1
    Gene Symbol VNN1
    Protein Tag IgG1 Fc Tag
    Protein Family Other Proteins
    Format Lyophilized

    Vanin 1 (VNN1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • This protein has a Fc region of human IgG1 at the C-terminus
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Activity: The activity of this protein has not been tested
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit