Learn More
SIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 216721AP
Catalog No. 50-173-3546
Add to cart
Description
Description
This gene encodes one of the proteins found in the SMN complex, which consists of several gemin proteins and the protein known as the survival of motor neuron protein. The SMN complex is localized to a subnuclear compartment called gems (gemini of coiled bodies) and is required for assembly of spliceosomal snRNPs and for pre-mRNA splicing. This protein interacts directly with the survival of motor neuron protein and it is required for formation of the SMN complex.
Specifications
Specifications
SIP1
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14893, Q9CQQ4, Q9QZP1
Component of gems 2, Gemin 2, GEMIN2, SIP1, SIP1 delta, SMN interacting protein 1
Rabbit
IgG
150 μL
-20°C
Primary
66603, 84404, 8487
Immunoprecipitation, Western Blot
Unconjugated
GEMIN2
Liquid
GEMIN2
SIP1 Fusion Protein Ag16500
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat