Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIPA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114461
Description
Positive controls that may be used include: Human Brain Tissue Lysate.
The product of this gene is a mitogen induced GTPase activating protein. It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.
Specifications
SIPA1 | |
Polyclonal | |
Unconjugated | |
SIPA1 | |
GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation associated gene 1; signal-induced proliferation-associated 1; signal-induced proliferation-associated gene 1; signal-induced proliferation-associated protein 1; SIPA1; Sipa-1; SPA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6494 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q96FS4 | |
SIPA1 | |
SIPA1 antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human SIPA1. The immunogen is located within amino acids 40 - 90 of SIPA1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction