Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sipper Systems for GENESYS 40–180 & BioMate 160 Spectrophotometers

Minimize sample handling and reduce the use of consumables

$486.50 - $3120.00


Products 3
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity & Availability  
14-385-360
View Documents
Thermo Scientific™
840296600
Sipper Pump with US Power Cable GENESYS 40–180 & BioMate 160 Spectrophotometers
Each for $1,875.00
Add to Cart
 
14-385-380
View Documents
Thermo Scientific™
840306900
Sipper System with US Power Cable GENESYS 140/150/180 & BioMate 160 Spectrophotometers
Each for $3,120.00
Add to Cart
 
14-385-394
View Documents
Thermo Scientific™
840307200
Sipper Sample Compartment GENESYS 140/150/180 & BioMate 160 Spectrophotometers
Each for $486.50
Add to Cart
 
Description

Description

Minimize sample handling and reduce the use of consumables with sipper sample holders. Integrated complete sipper system features:

  • Controlled by instrument on-board software
  • Powered by external AC/DC power supply unit
  • Sip by time or calibrate to sip by volume
  • Pre-programmed and custom sip sequences
  • Three pump speeds
  • Supports 1.6 mm wall thickness silicone tubing
  • C-Flex and Viton tubing materials are support in more limited range of sizes

Sipper systems available for select GENESYS™ and BioMate™ spectrophotometers, see ordering table for specific product compatibilities.

Includes:
  • Sipper pump with pump head
  • Sample compartment insert with cuvette holder and goose-neck aspiration tube guide
  • Quartz flow cell with connector tubing
  • Cable to connect to instrument
  • 30 cm of L/S 16 silicone tubing
  • Multi-voltage power supply & specified power cable
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.