Learn More
Sipper Systems for GENESYS 40–180 & BioMate 160 Spectrophotometers
Minimize sample handling and reduce the use of consumables
Manufacturer:  Thermo Scientific 840296600
 View more versions of this product
Catalog No. 14-385-360
Add to cart
Description
Description
Minimize sample handling and reduce the use of consumables with sipper sample holders. Integrated complete sipper system features:
  • Controlled by instrument on-board software
  • Powered by external AC/DC power supply unit
  • Sip by time or calibrate to sip by volume
  • Pre-programmed and custom sip sequences
  • Three pump speeds
  • Supports 1.6 mm wall thickness silicone tubing
  • C-Flex and Viton tubing materials are support in more limited range of sizes
Sipper systems available for select GENESYS™ and BioMate™ spectrophotometers, see ordering table for specific product compatibilities.
Includes:
  • Sipper pump with pump head
  • Sample compartment insert with cuvette holder and goose-neck aspiration tube guide
  • Quartz flow cell with connector tubing
  • Cable to connect to instrument
  • 30 cm of L/S 16 silicone tubing
  • Multi-voltage power supply & specified power cable
Specifications
Specifications
Sipper Pump with US Power Cable
GENESYS 40–180 & BioMate 160 Spectrophotometers