Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirius Red F3B, Spectrum™

Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
2610-10-8
C45H26N10Na6O21S6
1373.05
LDCKXVAPQFUIOI-ZFLDVXHKSA-H
hexasodium (3E)-4-oxo-7-({[(6E)-5-oxo-7-sulfonato-6-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-5,6-dihydronaphthalen-2-yl]carbamoyl}amino)-3-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2-sulfonate
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C3=CC=C(NC(=O)NC4=CC=C5C(=C4)C=C(\C(=N\NC4=CC=C(C=C4S([O-])(=O)=O)N=NC4=CC=C(C=C4)S([O-])(=O)=O)C5=O)S([O-])(=O)=O)C=C3C=C2S([O-])(=O)=O)C(=C1)S([O-])(=O)=O
2610-10-8
C45H26N10Na6O21S6
1373.05
LDCKXVAPQFUIOI-ZFLDVXHKSA-H
hexasodium (3E)-4-oxo-7-({[(6E)-5-oxo-7-sulfonato-6-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-5,6-dihydronaphthalen-2-yl]carbamoyl}amino)-3-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2-sulfonate
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C3=CC=C(NC(=O)NC4=CC=C5C(=C4)C=C(\C(=N\NC4=CC=C(C=C4S([O-])(=O)=O)N=NC4=CC=C(C=C4)S([O-])(=O)=O)C5=O)S([O-])(=O)=O)C=C3C=C2S([O-])(=O)=O)C(=C1)S([O-])(=O)=O

S1066, 2610-10-8, C45H26N10Na6O21S6

Spectrum™ Sirius Red F3B
CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Quantity 5 g