Learn More
Cell Signaling TechnologySupplier Diversity Partner SignalSilence(R) FoxO3a siRNA I (Mouse Specific) 300 ul DFS Item
Manufacturer:  Cell Signaling TechnologySupplier Diversity Partner 8620S
8620S , Forkhead box (Fox) proteins are a family of evolutionarily conserved transcription factors defined by the presence of a winged helix DNA binding domain called a Forkhead box (1). In humans, there are over 40 known Fox protein family members, divided into 19 subfamilies, which have evolved to regulate gene transcription in diverse and highly specialized biological contexts throughout development (2). Mutations that disrupt the expression of Fox gene family members have consequently been implicated in a broad array of human disorders, including immunological dysfunction, infertility, speech/language disorders, and cancer (3,4).
Catalog No. 50-194-896