Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP alpha Mouse anti-Human, Clone: SAF4.2, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen SIRP alpha
Clone SAF4.2
Concentration 0.5 mg/mL
Applications Flow Cytometry, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS2163100
View Documents
MilliporeSigma
MABS2163100UL
100 μL
Each for $387.00
Add to cart
 
MABS216325
View Documents
MilliporeSigma
MABS216325UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

SIRP alpha Monoclonal antibody specifically detects SIRP alpha in Human samples. It is validated for Flow Cytometry, Western Blotting.
Specifications

Specifications

SIRP alpha
0.5 mg/mL
Unconjugated
Mouse
Human
Human SIRP -Fc fusion protein consisting of the extracellular domain
Primary
Stable for 1 year at 2-8°C from date of receipt.
SAF4.2
Flow Cytometry, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit