Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP alpha Mouse anti-Human, Unconjugated, Clone: SAF10.1, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen SIRP alpha
Applications ELISA, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS21881MI
View Documents
MilliporeSigma
MABS2188100UG
100 μg
Each for $372.00
Add to cart
 
MABS21882MI
View Documents
MilliporeSigma
MABS218825UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

SIRP alpha Monoclonal antibody specifically detects SIRP alpha in Human samples. It is validated for ELISA, Western Blotting.
Specifications

Specifications

SIRP alpha
Unconjugated
Mouse
Tyrosine-protein phosphatase non-receptor type substrate 1;SHP substrate 1;SHPS-1;Brain Ig-like molecule with tyrosine-based activation motifs;Bit;CD172 antigen-like family member A;Inhibitory receptor SHPS-1;Macrophage fusion receptor;MyD-1 antigen;Signal-regulatory protein alpha-1;Sirp-alpha-1;Signal-regulatory protein alpha-2;Sirp-alpha-2;Signal-regulatory protein alpha-3;Sirp-alpha-3;p84;CD172a
Eukaryotically expressed SIRP-Fc fusion protein consisting of the extracellular domain.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Western Blot
Purified
Human
SIRPA, BIT, MFR, MYD1, PTPNS1, SHPS1, SIRP
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit