Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRP beta 1/gamma, Mouse anti-Human, Clone: OX117, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2810710.2MG
Description
SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation.Specifications
SIRP beta 1/gamma | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, ELISA, Immunoprecipitation | |
Mouse | |
Protein A purified | |
RUO | |
This antibody binds to human SIRP beta 1/gamma. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunoprecipitation | |
OX117 | |
Unconjugated | |
PBS | |
Recombinant human SIRPgamma:CD4 fusion protein. | |
0.2 mg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction