Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP beta 1/gamma, Mouse anti-Human, Clone: OX117, Novus Biologicals™

Mouse Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen SIRP beta 1/gamma
Clone OX117
Applications Flow Cytometry, ELISA, Immunoprecipitation
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015360
View Documents
Novus Biologicals
NBP2810710.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB015361
View Documents
Novus Biologicals
NBP2810710.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

SIRP beta 1/gamma Monoclonal antibody specifically detects SIRP beta 1/gamma in Human Samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation.
Specifications

Specifications

SIRP beta 1/gamma
Flow Cytometry, ELISA, Immunoprecipitation
Human
Primary
OX117
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit