Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP gamma/CD172g Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299880AF488

Catalog No. NB063332


Add to cart

Description

Description

SIRP gamma/CD172g Polyclonal specifically detects SIRP gamma/CD172g in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SIRP gamma/CD172g
Alexa Fluor 488
50mM Sodium Borate
bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma
Produced in rabbits immunized with purified, recombinant SIRP gamma/CD172g (NP_061026.2; Met 1-Ser 364).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Western Blot, ELISA
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Cell Cycle and Replication
Primary
55423
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit